Key Documents
Z377619
Sigma-Aldrich
Flow-Mesh gel and membrane support
sheet size 8 1/2 in. × 11 in., pkg of 20 sheets
Select a Size
1 PKG
$146.00
$146.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$146.00
About This Item
UNSPSC Code:
41105300
NACRES:
NB.51
$146.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 20 sheets
manufacturer/tradename
Diversified Biotech FM-100
sheet size
8 1/2 in. × 11 in.
General description
Inert perforated mesh with a high fraction of open volume, permitting free diffusion of solutions around gels. Use mesh to support gels and membranes for any solution treatment: staining/destaining, probing, incubation, etc. Supported gels and membranes will not tear or bend, and may be stacked for efficient use of time and solutions.
May be cut to size with scissors.
Legal Information
Flow-Mesh is a trademark of C.B.S. Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)