Skip to Content
Merck
All Photos(1)

Key Documents

Z358657

Millipore

Filter membranes, nitrocellulose

MF membrane, pore size 0.22 μm, diam. 293 mm

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$894.00

$894.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
25 EA
$894.00

About This Item

UNSPSC Code:
41104900
PubChem Substance ID:
NACRES:
NB.24

$894.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

MF membrane

manufacturer/tradename

Millipore GSWP29325

diam.

293 mm

pore size

0.22 μm

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert nitrocellulose membranes containing a small amount of cellulose acetate for improved handling. Autoclavable to 121°C; use below 75°C. Chemically compatible with dilute acids and bases, hydrocarbons, and non-polar liquids. MF-Millipore membranes have water extractables <5%, including a Triton wetting agent TF-Millipore membranes are Triton-free and have even lower water extractables than MF membranes. See Immobilon-NC in the Electrophoresis Equipment section for nitrocellulose blotting membranes.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinesh Chandra et al.
Clinical and experimental immunology, 154(2), 224-234 (2008-09-10)
The failure of Leishmania, an intracellular pathogen, to stimulate a pro-inflammatory response following entry into macrophages has been well reported. This occurs in spite of the fact that ligands for the toll-like receptors (TLR) have been recently shown on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service