Key Documents
Z331716
BRAND® macro pipette controller
green
Synonym(s):
brand pipette, macro pipette controller
Select a Size
1 EA
$258.00
$258.00
Estimated to ship onApril 16, 2025
Select a Size
Change View
1 EA
$258.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$258.00
Estimated to ship onApril 16, 2025
material
green
plastic (autoclavable)
silicone (autoclavable)
feature
autoclavable
manufacturer/tradename
BRAND 26201
volume
0.1-200 mL
General description
One single macro pipette controller covers the entire range of bulb and graduated pipettes from 0.1 to 200 mL. The conical silicone adapter offers a secure fit for the different diameters. The unit is fully autoclavable at 121°C (2 bar) according to DIN EN 285.
Supplied with a spare hydrophobic membrane filter which protects the system from liquid penetration.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)