Z327522
BRAND® graduated cylinder, PP, with blue printed scale or embossed scale
volume 10 mL, blue graduations
Select a Size
$138.00
Estimated to ship onMay 30, 2025
Select a Size
About This Item
$138.00
Estimated to ship onMay 30, 2025
Recommended Products
feature
subdivision 0.2 mL
manufacturer/tradename
BRAND
volume measuring range
10 mL
H
145 mm
color
blue graduations
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service