Documents
Z325368
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 20 mL, two marks
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
AR-Glas®
description
partial delivery
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29734
volume measuring range
20 mL, accuracy: 0.03 mL
capacity
20 mL
General description
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Other Notes
BR29734 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.