Key Documents
Z290653
Sigma-Aldrich
Vacuum filtration assembly funnel stems
glass membrane (support), membrane diam. 90 mm, connector for 40/35 joint (fritted glass support with side arm)
Synonym(s):
Filtration Funnel, Filtration Setup, Vacuum Filter Assembly
Select a Size
1 EA
$1,040.00
$1,040.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,040.00
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
$1,040.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
glass housing
glass membrane (support)
Joint
joint ST/NS40/35
packaging
pkg of 1 ea
membrane diam.
90 mm
fitting
connector for 40/35 joint (fritted glass support with side arm)
Choose from one of the most recent versions:
Certificates of Analysis (COA)