Skip to Content
Merck
All Photos(1)

Key Documents

Y1771

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without uracil, leucine, and tryptophan

Synonym(s):

Synthetic yeast drop-out mix, Yeast Synthetic Drop-out Media Supplements

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
$195.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
20 G
$195.00

About This Item

MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

form

powder

technique(s)

transformation: suitable

application(s)

microbiology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.
Yeast Synthetic Drop-out Medium Supplements, without uracil, leucine, and tryptophan has been used for screening:

  • the transformants of the PO1f strains which carried the pCRISPRyl_URA3sgRNA plasmids[1]
  • Yarrowia lipolytica PO1f strain positive transformants[2]
  • CRISPR- and RNA-assisted in vivo directed evolution (CRAIDE) mutants[3]

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Preparation Note

Dissolve 1.46 g of supplement per liter of medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Swietalski et al.
Yeast (Chichester, England), 39(3), 230-240 (2021-10-15)
Orotic acid (OA) is an intermediate of the pyrimidine biosynthesis with high industrial relevance due to its use as precursor for production of biochemical pyrimidines or its use as carrier molecule in drug formulations. It can be produced by fermentation
Emil D Jensen et al.
Bio-protocol, 12(5), e4346-e4346 (2022-05-21)
Directed evolution is a powerful approach to obtain genetically-encoded sought-for traits. Compared to the prolonged adaptation regimes to mutations occurring under natural selection, directed evolution unlocks rapid screening and selection of mutants with improved traits from vast mutated sequence spaces.
Paul Swietalski et al.
Microbial cell factories, 19(1), 100-100 (2020-05-13)
The secretory production of recombinant proteins in yeast simplifies isolation and purification but also faces possible complications due to the complexity of the secretory pathway. Therefore, correct folding, maturation and intracellular transport of the recombinant proteins are important processing steps

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service