Skip to Content
Merck
All Photos(3)

Key Documents

XNATS

Sigma-Aldrich

REDExtract-N-Amp Tissue PCR Kit

sufficient for 10 extractions, sufficient for 10 amplifications

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

usage

sufficient for 10 amplifications
sufficient for 10 extractions

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva. There is no need for mechanical disruption, organic extraction, column purification, or precipitation of the DNA. The kit includes a PCR ReadyMix, specially formulated for direct PCR amplification from the extract. This formulation uses an antibody-based hot start, for specific amplification. The REDExtract-N-Amp Tissue PCR Kit contains a tracking dye that allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp tissue PCR kit has been for:
  • DNA extraction
  • genotyping
  • polymerase chain reaction (PCR)

Features and Benefits

  • Perfect for quick genomic DNA isolation for genotyping
  • Single-step extraction of genomic DNA for PCR in 15 minutes
  • PCR ReadyMix, specially formulated for amplification directly from extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • Extract stable at 4oC for at least 6 months
  • REDExtract-N-Amp contains a tracking dye for convenient and direct loading of PCR reactions onto agarose gels for analysis

Components

Kits contain Tissue Preparation Solution, Extraction Solution, and REDExtract-N-Amp PCR Reaction Mix.

Other Notes

The REDExtract-N-Amp Tissue PCR Kits provide all the reagents necessary to rapidly extract DNA from a wide variety of cells and tissues and amplify targets of interest by PCR. A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR reaction mix for amplification directly from the extract. The REDExtract-N-Amp PCR ReadyMix contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reactions onto agarose gels for analysis.
For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Principle

The kit comes with validated protocols to extract and amplify genomic DNA from mouse-tails, hair, animal tissue, saliva, and buccal swabs. In a typical procedure, genomic DNA is extracted from a sample that has been incubated in the tissue preparation solution and extraction solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and REDExtract-N-Amp PCR ReadyMix, included in the kit.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Wang et al.
Nature communications, 9(1), 1544-1544 (2018-04-20)
Sexual dimorphism exists in energy balance, but the underlying mechanisms remain unclear. Here we show that the female mice have more pro-opiomelanocortin (POMC) neurons in the arcuate nucleus of hypothalamus than males, and female POMC neurons display higher neural activities
Bethany S Reid et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 239(4), 1188-1196 (2010-02-23)
Previous studies identified Inka1 as a gene regulated by AP-2alpha in the neural crest required for craniofacial morphogenesis in fish and frog. Here, we extend the analysis of Inka1 function and regulation to the mouse by generating a LacZ knock-in
Huan Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(26), 11942-11947 (2010-06-16)
During infection, vertebrates develop "sickness syndrome," characterized by fever, anorexia, behavioral withdrawal, acute-phase protein responses, and inflammation. These pathophysiological responses are mediated by cytokines, including TNF and IL-1, released during the innate immune response to invasion. Even in the absence
Farraj Albalawi et al.
Frontiers in cellular neuroscience, 11, 227-227 (2017-08-30)
Inflammatory responses play a key role in many neural pathologies, with localized signaling from the non-immune cells making critical contributions. The NLRP3 inflammasome is an important component of innate immune signaling and can link neural insult to chronic inflammation. The
Zhiyong Lei et al.
Frontiers in cardiovascular medicine, 8, 592362-592362 (2021-04-06)
Compromised cardiac function is a hallmark for heart failure, mostly appearing as decreased contractile capacity due to dysregulated calcium handling. Unfortunately, the underlying mechanism causing impaired calcium handling is still not fully understood. Previously the miR-132/212 family was identified as

Protocols

The REDExtract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

Extract-N-Amp kit protocol provides rapid DNA extraction yielding PCR-ready samples in 15 minutes with optimized PCR ReadyMix.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service