Recommended Products
matrix
4% cross-linked agarose
capacity
≥1.5 mg/mL binding capacity (uricase)
storage temp.
2-8°C
Application
Xanthine-agarose is used for protein chromatography, affinity chromatography and specialty resins. Xanthine-agarose has been used to purify and determine molecular properties of urate oxidase from Chlamydomonas reinhardtii. Xanthine-agarose has also been used to determine physicochemical properties and states of sulfhydryl groups of uricase from Candida utilis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
102.9 °F - closed cup
Flash Point(C)
39.4 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S S Mösli Waldhauser et al.
Phytochemistry, 45(7), 1407-1414 (1997-08-01)
Caffeine biosynthesis comprises sequential methylations at N-7, N-3 and N-1 of the xanthine ring catalysed by S-adenosyl-L-methionine (SAM)-dependent methyltransferase activities that, to date, have not been resolved. Enzyme extracts were prepared from young, emerging coffee leaflets and following anion exchange
Débora da Silva Freitas et al.
International journal of pharmaceutics, 387(1-2), 215-222 (2009-12-09)
PEGylation is a successful strategy for improving the biochemical and biopharmaceutical properties of proteins and peptides through the covalent attachment of polyethylene glycol chains. In this work, purified recombinant uricase from Candida sp. (UC-r) was modified by PEGylation with metoxypolyethilenoglycol-p-nitrophenyl-carbonate
Isolation and characterization of uricase from bean leaves and its comparison with uredospore enzymes.
Montalbini, P., et al.
Plant Science, 147(2), 139-147 (1999)
Marialaura Marchetti et al.
Scientific reports, 6, 38302-38302 (2016-12-07)
Urate oxidase (Uox) catalyses the first reaction of oxidative uricolysis, a three-step enzymatic pathway that allows some animals to eliminate purine nitrogen through a water-soluble compound. Inactivation of the pathway in hominoids leads to elevated levels of sparingly soluble urate
Uricase from leaves: its purification and characterization from three different higher plants.
Montalbini, P., et al.
Planta, 202(3), 277-283 (1997)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service