X1000
Xanthosine 5′-monophosphate disodium salt
≥85%
Synonym(s):
XMP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H11N4Na2O9P
CAS Number:
Molecular Weight:
408.17
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Recommended Products
Assay
≥85%
storage temp.
−20°C
SMILES string
[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)n2cnc3C(=O)NC(=O)Nc23
InChI
1S/C10H13N4O9P.2Na/c15-5-3(1-22-24(19,20)21)23-9(6(5)16)14-2-11-4-7(14)12-10(18)13-8(4)17;;/h2-3,5-6,9,15-16H,1H2,(H2,19,20,21)(H2,12,13,17,18);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
InChI key
QQPVYRBCIRHGSZ-LGVAUZIVSA-L
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Diego Alberto Montano Vizcarra et al.
Acta histochemica, 122(2), 151484-151484 (2020-01-07)
The aim of this study was to evaluate whether the addition of synthetic polymers to the vitrification solution affected follicular morphology and development and the expression of Ki-67, Aquaporin 3 (AQP3) and cleaved Caspase-3 proteins in ovarian tissue of the
Zaipul I Md Dom et al.
British journal of clinical pharmacology, 84(10), 2433-2442 (2018-07-06)
Although therapeutic drug monitoring of plasma mycophenolic acid (MPA) concentrations has been recommended to individualize dosage in transplant recipients, little is known regarding lymphocyte concentrations of MPA, where MPA inhibits inosine monophosphate dehydrogenase (IMPDH). This study investigated the utility of
G I Mohammed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 229, 117929-117929 (2020-01-10)
The current study explores the use of quercetin for developing a highly selective spectrofluorimetric methodology for trace determination, speciation and thermodynamic characterization of tungstate (WO42-) species in water. The study relies on the principle of chelate formation between WO42- and
Jana Enderes et al.
Frontiers in immunology, 11, 581111-581111 (2021-02-02)
Postoperative ileus (POI) is triggered by an innate immune response in the muscularis externa (ME) and is accompanied by bacterial translocation. Bacteria can trigger an innate immune response via toll-like receptor (TLR) activation, but the latter's contribution to POI has
Gero Weißbarth et al.
Therapeutic drug monitoring, 42(2), 282-288 (2019-08-20)
Mycophenolic acid (MPA), a powerful inhibitor of lymphocyte proliferation, is widely used in transplantation medicine and as a glucocorticoid-sparing agent in rheumatic and inflammatory diseases. As inosine-5'-monophosphate dehydrogenase (IMPDH), the target enzyme of MPA, shows high interindividual variability in its
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service