Skip to Content
Merck
All Photos(1)

Documents

WPB01558

Whirl-Pak® Black Bag

capacity 1627 mL (17 oz), W × L × thickness 19 cm × 30 cm × 0.064 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

material

black linear low-density polyethylene (LLDPE)

sterility

sterile

feature

label plain white

packaging

pkg of 1000 ea (2 boxes of 500EA)

manufacturer/tradename

Nasco Whirl-pak® B01558

W × L × thickness

19 cm × 30 cm × 0.064 mm

capacity

1627 mL (17 oz)

Looking for similar products? Visit Product Comparison Guide

General description

When it is necessary to protect a sample from light, the Whirl-Pak black bag is specially designed for this application. The polyethylene film is made from pigmented resins so the black color is an integral part of the bag and not a printed layer. The result is an opaque bag providing maximum protection for samples that are sensitive to light. It is ideal for pharmaceuticals, seasonings, flavors, food supplements, vitamins, marine life, and many other liquid or solid products. It is FDA approved for food contact.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.

Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.

Features and Benefits

  • Opaque to protect light sensitive contents
  • FDA approved
  • Built-in closure for secure sample collection of content sensitive to light.

Legal Information

Whirl-pak is a registered trademark of The Aristotle Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service