Skip to Content
Merck
All Photos(2)

Documents

WH0387129M1

Sigma-Aldrich

Monoclonal Anti-GPR154 antibody produced in mouse

clone 2F5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-G protein-coupled receptor 154, Anti-GPRA, Anti-PGR14, Anti-VRR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPSR1(387129)

Related Categories

General description

This gene is a member of the G protein-coupled receptor 1 family and encodes a plasma membrane protein. Increased expression of this gene in ciliated cells of the respiratory epithelium and in bronchial smooth muscle cells is associated with asthma. Mutations in this gene have also been associated with this disease. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. (provided by RefSeq)

Immunogen

GPR154 (NP_997056, 2 a.a. ~ 53 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PANFTEGSFDSSGTGQTLDSSPVACTETVTFTEVVEGKEWGSFYYSFKTEQL

Biochem/physiol Actions

G protein-coupled receptor 154 (GPR154) is involved in processes like nociception and inflammation. Polymorphisms in the gene encoding it are linked to asthma and chronic inflammatory diseases.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kariina Laas et al.
Journal of psychopharmacology (Oxford, England), 29(8), 878-883 (2015-03-07)
Administration of neuropeptide S (NPS) elicits anxiolysis, arousal and higher activity in rodents. In humans, the NPS receptor (NPSR1) gene rs324981 A/T (Asn(107)Ile) polymorphism is associated with fear responses and anxiety. We have recently revealed an association of NPSR1 with
M Henström et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 26(10), 1417-1425 (2014-08-06)
Recurrent abdominal pain (RAP) occurs frequently among children and is one of the cardinal symptoms of functional gastrointestinal disorders (FGID). The mechanisms of visceral pain and RAP are not fully understood. A heritable component has been demonstrated and a few

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service