Skip to Content
Merck
All Photos(6)

Documents

WH0083667M3

Sigma-Aldrich

Monoclonal Anti-SESN2 antibody produced in mouse

clone 3B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp761M0212, Anti-DKFZp761M02121, Anti-HI95, Anti-SES2, Anti-SEST2, Anti-sestrin 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SESN2(83667)

General description

This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions. (provided by RefSeq)

Immunogen

SESN2 (AAH13304.1, 1 a.a. ~ 480 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MIVADSECRAELKDYLRFAPGGVGDSGPGEEQRESRARRGPRGPSAFIPVEEVLREGAESLEQHLGLEALMSSGRVDNLAVVMGLHPDYFTSFWRLHYLLLHTDGPLASSWRHYIAIMAAARHQCSYLVGSHMAEFLQTGGDPEWLLGLHRAPEKLRKLSEINKLLAHRPWLITKEHIQALLKTGEHTWSLAELIQALVLLTHCHSLSSFVFGCGILPEGDADGSPAPQAPTPPSEQSSPPSRDPLNNSGGFESARDVEALMERMQQLQESLLRDEGTSQEEMESRFELEKSESLLVTPSADILEPSPHPDMLCFVEDPTFGYEDFTRRGAQAPPTFRAQDYTWEDHGYSLIQRLYPEGGQLLDEKFQAAYSLTYNTIAMHSGVDTSVLRRAIWNYIHCVFGIRYDDYDYGEVNQLLERNLKVYIKTVACYPEKTTRRMYNLFWRHFRHSEKVHVNLLLLEARMQAALLYALRAITRYMT

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana Saveljeva et al.
Oncotarget, 7(11), 12254-12266 (2016-03-02)
Upregulation of SESTRIN 2 (SESN2) has been reported in response to diverse cellular stresses. In this study we demonstrate SESTRIN 2 induction following endoplasmic reticulum (ER) stress. ER stress-induced increases in SESTRIN 2 expression were dependent on both PERK and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service