Skip to Content
Merck
All Photos(3)

Key Documents

WH0051773M5

Sigma-Aldrich

Monoclonal Anti-HBXAP antibody produced in mouse

clone 3E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-RSF1, Anti-XAP8, Anti-hepatitis B virus x associated protein, Anti-p325

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RSF1(51773)

Related Categories

General description

HBXAP is involved in transcription repression, transcription coactivation when associated with hepatitis B virus X protein (HBX), and chromatin remodeling and spacing when associated with SNF2H (MIM 603375).[supplied by OMIM

Immunogen

HBXAP (NP_057662.3, 1343 a.a. ~ 1441 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IESDEEEDFENVGKVGSPLDYSLVDLPSTNGQSPGKAIENLIGKPTEKSQTPKDNSTASASLASNGTSGGQEAGAPEEEEDELLRVTDLVDYVCNSEQL

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service