Skip to Content
Merck
All Photos(2)

Documents

WH0023569M1

Sigma-Aldrich

Monoclonal Anti-PADI4 antibody produced in mouse

clone 4D8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PAD, Anti-PADI5, Anti-PDI4, Anti-PDI5, Anti-peptidyl arginine deiminase, type IV

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PADI4(23569)

General description

Peptidyl arginine deiminase 4 (PADI4) is one of the four PADIs found in humans. The protein consists of 663 amino acids. PADI4 gene is localized to human chromosome 1p36. It is expressed in hematopoietic tissues, such as spleen, thymus, peripheral blood leukocytes, fetal liver and bone marrow.

Immunogen

PADI4 (NP_004247, 2 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
AQGTLIRVTPEQPTHAVCVLGTLTQLDICSSAPEDCTSFSINASPGVVVDIAHSPPAKKKSTGSSTWPLDPGVEVTLTMKAASGSTGDQKVQISYYGPKTPPVKALLYL

Biochem/physiol Actions

Peptidyl arginine deiminase 4 (PADI4) is responsible for the post-translational conversion of peptidylarginine to citrulline, in the presence of calcium ions. In individuals with rheumatoid arthritis (RA), this gene is expressed in hematological cells and synovial tissues, and variant in this gene is linked with susceptibility to RA. The expression of this protein is linked with DNA hypermethylation in acute promyelocytic leukemia (APL).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional haplotypes of PADI4, encoding citrullinating enzyme peptidylarginine deiminase 4, are associated with rheumatoid arthritis.
Suzuki A, et.al
Nature Genetics, 34(4), 395-402 (2003)
Localization of peptidylarginine deiminase 4 (PADI4) and citrullinated protein in synovial tissue of rheumatoid arthritis.
Chang X, et.al
Rheumatology (Oxford, England), 44(1), 40-50 (2005)
A novel PAD4/SOX4/PU.1 signaling pathway is involved in the committed differentiation of acute promyelocytic leukemia cells into granulocytic cells.
Song G, et.al
Oncotarget, 7(3), 3144-3157 (2016)
Localization of peptidylarginine deiminase 4 (PADI4) and citrullinated protein in synovial tissue of rheumatoid arthritis.
Chang X et al
Rheumatology (Oxford, England), 44(1), 40-50 (2005)
A novel PAD4/SOX4/PU.1 signaling pathway is involved in the committed differentiation of acute promyelocytic leukemia cells into granulocytic cells.
Song G et al
Oncotarget, 7(3), 3144-3157 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service