Skip to Content
Merck
All Photos(3)

Key Documents

WH0010247M1

Sigma-Aldrich

Monoclonal Anti-HRSP12 antibody produced in mouse

clone 2B8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-P14.5, Anti-PSP, Anti-UK114, Anti-heat-responsive protein 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HRSP12(10247)

General description

HRSP12 (Heat-responsive protein 12) is a trichloroacetic acid soluble protein belonging to the hypothetical family of small proteins, termed as YER057c/YJGF family. It has a molecular weight of 14.5kDa. It is expressed in monocytes, liver and kidney tumor cells. Its high level of expression has been reported in fully differentiated monocytes.

Immunogen

HRSP12 (AAH10280.1, 1 a.a. ~ 137 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSSLIRRVISTAKAPGAIGPYSQAVLVDRTIYISGQIGMDPSSGQLVSGGVAEEAKQALKNMGEILKAAGCDFTNVVKTTVLLADINDFNTVNEIYKQYFKSNFPARAAYQVAALPKGSRIEIEAVAIQGPLTTASL

Biochem/physiol Actions

HRSP12 (Heat-responsive protein 12) mainly acts as a translational inhibitor protein. It has been proposed that the protein may have the potentiality to perform in various cellular mechanisms such as a differentiation-dependent regulation of protein synthesis in hepatocytes, renal tubular epithelial cells, smooth muscle cells and mononuclear phagocytes (MNP).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of Homo sapiens protein hp14.5.
Babu A Manjasetty et al.
Proteins, 54(4), 797-800 (2004-03-05)
G Schmiedeknecht et al.
European journal of biochemistry, 242(2), 339-351 (1996-12-01)
A trichloroacetic-acid-soluble 14.5-kDa protein (p14.5) has been isolated from human mononuclear phagocytes (MNP) by a combination of trichloroacetic acid extraction, preparative electrophoresis and hydrophobic affinity chromatography; five tryptic peptides were subjected to protein sequencing. The full-length cDNA of the protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service