Skip to Content
Merck
All Photos(5)

Documents

WH0007855M1

Sigma-Aldrich

Monoclonal Anti-FZD5 antibody produced in mouse

clone 6A3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HFZ5, Anti-frizzled homolog 5 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6A3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD5(7855)

General description

The frizzled class receptor 5 (FZD5) gene encodes a receptor FZ5 for Wnt proteins and it is localized on human chromosome 2q33.3.

Immunogen

FZD5 (ENSP00000354607, 72 a.a. ~ 161 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PLVEIQCSPDLRFFLCSMYTPICLPDYHKPLPPCRSVCERAKAGCSPLMRQYGFAWPERMSCDRLPVLGRDAEVLCMDYNRSEATTAPPR

Biochem/physiol Actions

Frizzled class receptor 5 (FZD5) is a receptor component for the canonical and non-canonical Wnt signaling pathways, which is necessary for cell proliferation and cell differentiation. It might serve as an important biomarker for eye malformation. The Wnt5a-FZD5 complex is known to support the functions of innate immunity. Fzd5, along with GCM1 (chorion-specific transcription factor), is involved in a feedback mechanism that regulates trophoblast differentiation and chorionic branching in placenta.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wnt5a-Rac1-NF-?B homeostatic circuitry sustains innate immune functions in macrophages.
Naskar D
Journal of Immunology (2014)
A positive feedback loop involving Gcm1 and Fzd5 directs chorionic branching morphogenesis in the placenta.
Lu J
PLoS Biology (2013)
A secreted WNT-ligand-binding domain of FZD5 generated by a frameshift mutation causes autosomal dominant coloboma.
Liu C
Human Molecular Genetics (2016)
Strong linkage on 2q33.3 to familial early-onset generalized osteoarthritis and a consideration of two positional candidate genes.
Meulenbelt I
European Journal of Human Genetics (2006)
Functional analysis of dishevelled-3 phosphorylation identifies distinct mechanisms driven by casein kinase 1? and frizzled5.
Bernatik O
The Journal of Biological Chemistry (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service