Skip to Content
Merck
All Photos(7)

Documents

WH0007004M1

Sigma-Aldrich

Monoclonal Anti-TEAD4 antibody produced in mouse

clone 5H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-EFTR2, Anti-MGC9014, Anti-RTEF1, Anti-TCF13L1, Anti-TEA domain family member 4, Anti-TEF3, Anti-TEFR1, Anti-hRTEF1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H3, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEAD4(7004)

General description

This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene. (provided by RefSeq)

Immunogen

TEAD4 (NP_003204, 151 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ARGPGRPAVSGFWQGALPGQAGTSHDVKPFSQQTYAVQPPLPLPGFESPAGPAPSPSAPPAPPWQGRSVASSKLWMLEFSAFLEQQQDPDTYNKHLFVHIGQSSPSYSDP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong-Yu Wang et al.
Chinese journal of cancer, 35, 47-47 (2016-05-22)
Osteosarcoma is the most common bone malignancy in children and adolescents, and 20%-30% of the patients suffer from poor prognosis because of individual chemoresistance. The Hippo/yes-associated protein (YAP) signaling pathway has been shown to play a role in tumor chemoresistance
Qinghe Song et al.
PloS one, 10(3), e0120790-e0120790 (2015-03-27)
The Yes-associated protein (YAP), a transcriptional coactivator inactivated by the Hippo tumor suppressor pathway, functions as an oncoprotein in a variety of cancers. However, its contribution to breast cancer remains controversial. This study investigated the role of YAP in breast
Ting Wang et al.
Biochimica et biophysica acta. Molecular basis of disease, 1864(5 Pt A), 1744-1753 (2018-03-03)
The transcriptional co-activator Yes-associated protein (YAP) has been implicated as an oncogene and is found to promote breast cancer metastasis. However, the pro-metastatic mechanism of YAP remains unclear. Here, we demonstrated that YAP functions as a transcriptional repressor of growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service