Skip to Content
Merck
All Photos(2)

Key Documents

WH0002548M1

Sigma-Aldrich

Monoclonal Anti-GAA antibody produced in mouse

clone 3C6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-LYAG, Anti-glucosidase, α acid (Pompe disease, glycogen storage disease type II)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAA(2548)

General description

Lysosomal α-glucosidase (GAA), a glycoprotein and member of glycoside hydrolase family GH31, comprises a trefoil type-P domain, catalytic GH31 domain, distal, proximal, and an N-terminal β-sheet domain. The GAA gene is mapped to human chromosome 17q25.3. It undergoes various proteolytical and N-glycan processing in the late endosomal/lysosomal compartment to become an active form.

Immunogen

GAA (AAH40431, 851 a.a. ~ 952 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GEARGELFWDDGESLEVLERGAYTQVIFLARNNTIVNELVRVTSEGAGLQLQKVTVLGVATAPQQVLSNGVPVSNFTYSPDTKVLDICVSLLMGEQFLVSWC

Biochem/physiol Actions

Lysosomal α-glucosidase (GAA) uses mannose-6-phosphate receptors for its localization on the lysosomes. It mediates the hydrolysis of glycogen to glucose. Mutations in the GAA gene impair acid alpha-glucosidase enzyme activity. Deficiency of GAA leads to a rare lysosomal storage disease namely Pompe disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hossein Moravej et al.
Iranian journal of medical sciences, 43(2), 218-222 (2018-05-12)
Pompe disease (PD), also known as "glycogen storage disease type II (OMIM # 232300)" is a rare autosomal recessive disorder characterized by progressive glycogen accumulation in cellular lysosomes. It ultimately leads to cellular damage. Infantile-onset Pompe disease (IOPD) is the
Lysosomal acid alpha-glucosidase consists of four different peptides processed from a single chain precursor.
Moreland, et al.
The Journal of Biological Chemistry, 280, 6780-6791 (2021)
Véronique Roig-Zamboni et al.
Nature communications, 8(1), 1111-1111 (2017-10-25)
Pompe disease, a rare lysosomal storage disease caused by deficiency of the lysosomal acid α-glucosidase (GAA), is characterized by glycogen accumulation, triggering severe secondary cellular damage and resulting in progressive motor handicap and premature death. Numerous disease-causing mutations in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service