Skip to Content
Merck
All Photos(8)

Key Documents

WH0001021M1

Sigma-Aldrich

Monoclonal Anti-CDK6 antibody produced in mouse

clone 8H4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC59692, Anti-PLSTIRE, Anti-cyclin-dependent kinase 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8H4, monoclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK6(1021)

General description

Cyclin dependent kinase 6 (CDK6) is encoded by the gene mapped to human chromosome 7q21.2. The encoded protein is ubiquitously expressed and is a member of CDK family.
The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This kinase is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression and G1/S transition. The activity of this kinase first appears in mid-G1 phase, which is controlled by the regulatory subunits including D-type cyclins and members of INK4 family of CDK inhibitors. This kinase, as well as CDK4, has been shown to phosphorylate, and thus regulate the activity of, tumor suppressor protein Rb. (provided by RefSeq)

Immunogen

CDK6 (NP_001250, 3 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KDGLCRADQQYECVAEIGEGAYGKVFKARDLKNGGRFVALKRVRVQTGEEGMPLSTIREVAVLRHLETFEHPNVVRLFDVCTVSRTDRETKLTLVFE

Biochem/physiol Actions

Cyclin dependent kinase 6 (CDK6) plays a vital role in regulation of cell‐cycle and thus, it is considered as a potent therapeutic target for cancers.The protein expressed in human hematopoietic stem cells (HSC), modulates quiescence exit. Elevated expression of the gene has been observed in various types of cancers such as leukemia and lymphomas.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palbociclib can overcome mutations in cyclin dependent kinase 6 that break hydrogen bonds between the drug and the protein
Hernandez Maganhi S
Protein Science, 26, 870-879 (2017)
A Kinase-Independent Function of CDK6 Links the Cell Cycle to Tumor Angiogenesis
Kollmann K
Cancer Cell, 30, 359-360 (2016)
MLL fusion-driven activation of CDK6 potentiates proliferation in MLL-rearranged infant ALL
van der Linden MH
Cell Cycle, 13, 834-844 (2014)
CDK6 Levels Regulate Quiescence Exit in Human Hematopoietic Stem Cells
Laurenti E
Cell Stem Cell, 16, 302-313 (2015)
Ma Yanchun et al.
European journal of pharmacology, 851, 43-51 (2019-02-20)
Triptolide, the component of traditional Chinese herb, has been used as an inflammatory medicine and reported to be anti-tumor for various cancers recently. However, the effect of triptolide on Esophageal Squamous Cell Cancer (ESCC) has not yet been elucidated. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service