Skip to Content
Merck
All Photos(2)

Documents

WH0000675M1

Sigma-Aldrich

Monoclonal Anti-BRCA2 antibody produced in mouse

clone 3D12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FACD, Anti-FAD, Anti-FAD1, Anti-FANCB, Anti-FANCD, Anti-FANCD1, Anti-breast cancer 2, early onset

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRCA2(675)

General description

Breast cancer type 2 susceptibility protein (BRCA2), DNA repair associated gene is located on human chromosome 13q13.1. BRCA2 and BRCA1 colocalize in subnuclear foci in somatic cells and on the axial elements of developing synaptonemal complexes.

Immunogen

BRCA2 (NP_000050, 3319 a.a. ~ 3418 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SPQMTPFKKFNEISLLESNSIADEELALINTQALLSGSTGEKQFISVSESTRTAPTSSEDYLRLKRRCTTSLIKEQESSQASTEECEKNKQDTITTKKYI

Biochem/physiol Actions

Breast cancer type 2 susceptibility protein (BRCA2), DNA repair associated protein encodes a protein that promotes homologous recombination, which is essential for normal development. Mutations causes tumor formation and cell lethality. Bi-allelic mutations of BRCA2 is associated with Fanconi anemia (FA). BRCA2 alterations can be used as a prognostic marker for pancreatic cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of BRCA2 mutation status as diagnostic, predictive, and prognosis biomarker for pancreatic cancer
Martinez-Useros J and Garcia-Foncillas J
BioMed Research International, 2016, e2875-e2875 (2016)
Stable interaction between the products of the BRCA1 and BRCA2 tumor suppressor genes in mitotic and meiotic cells
Chen J, et al.
Molecular Cell, 2(3), 317-328 (1998)
Distinct genomic profiles in hereditary breast tumors identified by array-based comparative genomic hybridization
Jonsson G, et al.
Cancer research, 65(17), 7612-7621 (2005)
Acquired cross-linker resistance associated with a novel spliced BRCA2 protein variant for molecular phenotyping of BRCA2 disruption
Meyer S, et al.
Cell Death & Disease, 8(6), e2875-e2875 (2017)
BRCA2 suppresses replication stress-induced mitotic and G1 abnormalities through homologous recombination.
Feng W and Jasin M
Nature Communications, 8(1), 525-525 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service