Skip to Content
Merck
All Photos(1)

Documents

W4639

Sigma-Aldrich

Wash Solution 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

storage temp.

room temp

Related Categories

General description

This product is used as a column wash after DNA binding performed with GenElute kits.

Preparation Note

For usage requirements, please refer to the kit procedure. Ethanol is required as part of the Wash Solution 2 preparation.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service