Skip to Content
Merck
All Photos(1)

Key Documents

W3020

Sigma-Aldrich

WKYMVdM trifluoroacetate salt

≥98% (HPLC), powder

Synonym(s):

Trp-Lys-Tyr-Met-Val-D-Met NH2 trifluoroacetate salt trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$304.00

$304.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$304.00

About This Item

Empirical Formula (Hill Notation):
C41H61N9O7S2 · xC2HF3O2
CAS Number:
Molecular Weight:
856.11 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$304.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: >2 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCSC)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)Cc2c[nH]c3ccccc23)C(C)C)C(N)=O

InChI

1S/C41H61N9O7S2.C2HF3O2/c1-24(2)35(41(57)46-31(36(44)52)16-19-58-3)50-39(55)33(17-20-59-4)48-40(56)34(21-25-12-14-27(51)15-13-25)49-38(54)32(11-7-8-18-42)47-37(53)29(43)22-26-23-45-30-10-6-5-9-28(26)30;3-2(4,5)1(6)7/h5-6,9-10,12-15,23-24,29,31-35,45,51H,7-8,11,16-22,42-43H2,1-4H3,(H2,44,52)(H,46,57)(H,47,53)(H,48,56)(H,49,54)(H,50,55);(H,6,7)/t29-,31+,32-,33-,34-,35-;/m0./s1

InChI key

MYVYQWVNXDVTBW-HQORXVKSSA-N

Amino Acid Sequence

Trp-Lys-Tyr-Met-Val-D-Met NH2

Biochem/physiol Actions

WKYMVm is a very potent agonist of formyl peptide receptor members FPR1/FPR2 (EC50 1 nM) and FPRL1/FPRL2 (75 pM and 3 nM, respectively). WKYMVm stimulation of leukocytes has been shown to elevate intracellular calcium, induce chemotaxis, superoxide generation, cell killing and phagocytosis.
WKYMVm is a very potent agonist of formyl peptide receptor members FPR1/FPR2 (EC50 1 nM) and FPRL1/FPRL2 (75pM and 3 nM, respectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service