Skip to Content
Merck
All Photos(1)

Key Documents

W0641

Sigma-Aldrich

W-84 dibromide

≥98% (HPLC), solid

Synonym(s):

Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H44Br2N4O4
CAS Number:
Molecular Weight:
708.52
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ~11 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

[Br-].[Br-].C[N+](C)(CCCCCC[N+](C)(C)CCCN1C(=O)c2ccccc2C1=O)CCCN3C(=O)c4ccccc4C3=O

InChI

1S/C32H44N4O4.2BrH/c1-35(2,23-13-19-33-29(37)25-15-7-8-16-26(25)30(33)38)21-11-5-6-12-22-36(3,4)24-14-20-34-31(39)27-17-9-10-18-28(27)32(34)40;;/h7-10,15-18H,5-6,11-14,19-24H2,1-4H3;2*1H/q+2;;/p-2

InChI key

DZRJZDQAGMZGGA-UHFFFAOYSA-L

Biochem/physiol Actions

Potent allosteric modulator of M2 muscarinic acetylcholine receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Ping Huang et al.
Molecular pharmacology, 68(3), 769-778 (2005-06-07)
The structurally divergent agents gallamine and hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide (W84) are known to interact competitively at a common allosteric site on muscarinic receptors. Previous studies reported that the M2 selectivity of gallamine depended largely on the EDGE (172-175) sequence in the second
Marion Mohr et al.
Journal of medicinal chemistry, 47(12), 3324-3327 (2004-05-28)
Various fragments of the hexamethonio-type allosteric agent W84 were linked to the secondary amino group of the muscarinic M(2) acetylcholine receptor-preferring antagonist AF-DX 384 to increase the area of attachment with the allosteric site. Addition of only the phthalimido moiety
Ulrike Holzgrabe et al.
Journal of molecular neuroscience : MN, 30(1-2), 165-168 (2006-12-29)
Allosteric modulators of ligand-receptor interactions are found for a variety of receptors (Christopoulos, 2002). Allosteric agents attach to a binding site being topographically distinct from the site for conventional (orthosteric) agonists or antagonists. In the case of the muscarinic receptor
K Mohr et al.
Pharmacology & toxicology, 75(6), 391-394 (1994-12-01)
The bis-quaternary W84, hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)-ammonium bromide], is a potent allosteric modulator of M2-cholinoceptors. In this study we aimed at quantifying its allosteric effect on the dissociation of [3H]pirenzepine from M1-cholinoceptors in rat cerebral cortex and to measure the effects on association
C Tränkle et al.
Molecular pharmacology, 53(2), 304-312 (1998-03-14)
The hypothesis was tested that M2-selective antagonists partially utilize the allosteric site of muscarinic M2 receptors. The interactions of the allosteric agent W84 (hexane-1, 6-bis[dimethyl-3'-phthalimidopropyl-ammonium bromide]) were studied with the M2/M4-selective AF-DX 384 [(+/-)-5, 11-dihydro-11-([(2-(2-[(dipropylamino)methyl]-1-piperidinyl)ethyl)amino]carbonyl)-6H-pyrido(2,3-b)(1,4)-benzodiazepine-6-one], the nonselective N-methylscopolamine (NMS), and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service