Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 4 x 1 ea | Check Cart for Availability | $477.00 |
About This Item
NACRES:
NB.24
UNSPSC Code:
41104923
Size:
96 wells
$477.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Quality Segment
size
96 wells
Application
The Filter Plate can be used with Sigma′s GenElute 96 Well Total RNA Purification Kit to remove cell debris and prevent the RNA Binding Plate from clogging. The Filter Plate is used to filter the lysate before applying the lysate to the RNA Binding Plate (GenElute 96 Well Total RNA Purification Kit).
1 of 1
This Item | |||
|---|---|---|---|
| size 96 wells | size - | size - | size - |
| Quality Level 100 | Quality Level - | Quality Level 400 | Quality Level 400 |
Choose from one of the most recent versions:
Already Own This Product?
Global Trade Item Number
| SKU | GTIN |
|---|---|
| VM03-4X1EA | 04061837481680 |