Skip to Content
Merck
All Photos(1)

Key Documents

V5130

Sigma-Aldrich

Val-His-Leu-Thr-Pro-Val-Glu-Lys acetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Other Notes

Amino-terminal octapeptide of the HbS (sickle-cell hemoglobin) β-chain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene mutations in human haemoglobin: the chemical difference between normal and sickle cell haemoglobin.
V M INGRAM
Nature, 180(4581), 326-328 (1957-08-17)
A Eastlake et al.
The Journal of biological chemistry, 251(20), 6426-6430 (1976-10-25)
Five peptides from the NH2-terminal region of the beta chain of hemoglobin S, betaS (1-13), betaS (3-13), betaS (1-8), betaS (4-10), And betaS (4-8), have been synthesized by a rapid solid phase method based on the Merrifield procedure. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service