Skip to Content
Merck
All Photos(1)

Key Documents

V4890

Sigma-Aldrich

Vindesine sulfate salt hydrate

≥95% (HPLC)

Synonym(s):

3-(Aminocarbonyl)-O4-deacetyl-3-de(methoxycarbonyl)vincaleukoblastine sulfate salt hydrate, Desacetylvinblastine amide sulfate salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$154.00
25 MG
$596.00

$154.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$154.00
25 MG
$596.00

About This Item

Empirical Formula (Hill Notation):
C43H55N5O7 · H2SO4 · xH2O
CAS Number:
Molecular Weight:
852.00 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$154.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

off-white to light yellow

solubility

deionized water: ≥50 mg/mL

originator

Eli Lilly

storage temp.

−20°C

SMILES string

O.OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@H]7[C@](O)([C@H](O)[C@]8(CC)C=CC[N@H]9CC[C@]67[C@H]89)C(N)=O)C1

InChI

1S/C43H55N5O7.H2O4S.H2O/c1-6-39(52)21-25-22-42(38(51)55-5,33-27(13-17-47(23-25)24-39)26-11-8-9-12-30(26)45-33)29-19-28-31(20-32(29)54-4)46(3)35-41(28)15-18-48-16-10-14-40(7-2,34(41)48)36(49)43(35,53)37(44)50;1-5(2,3)4;/h8-12,14,19-20,25,34-36,45,49,52-53H,6-7,13,15-18,21-24H2,1-5H3,(H2,44,50);(H2,1,2,3,4);1H2/t25-,34+,35-,36-,39+,40-,41-,42+,43+;;/m1../s1

InChI key

TUDLKTILIBRXNU-RIHOAIHNSA-N

Biochem/physiol Actions

Microtubule inhibitor
Microtubule inhibitor. Vindesine binds to and stabilizes tubulin, thereby preventing tubulin polymerization, preventing mitosis, and arresting cells in metaphase.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intensified chemotherapy for diffuse large B-cell lymphomas.
Julie M Vose
Lancet (London, England), 378(9806), 1828-1829 (2011-11-29)
Olivier Fitoussi et al.
Haematologica, 96(8), 1136-1143 (2011-05-07)
As rituximab combined with CHOP improves complete remission and overall survival in diffuse large B-cell lymphoma, intensified chemotherapy followed by autologous stem-cell transplantation has also been advocated for high-risk patients. The aim of this study was to establish whether or
Yukitaka Nakano et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 132(6), 727-732 (2012-06-13)
This paper reports a specific and sensitive enzyme-linked immunosorbent assay (ELISA) for pharmacokinetic studies of vindesine (VDS). Anti-VDS antibody was obtained by immunizing rabbits with VDS conjugated with bovine serum albumin using N-[β-(4-diazophenyl) ethyl] maleimide as a heterobifunctional coupling agent.
Teodoro Chisesi et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 29(32), 4227-4233 (2011-10-13)
The Intergruppo Italiano Linfomi HD9601 trial compared doxorubicin, bleomycin, vinblastine, and dacarbazine (ABVD) versus doxorubicin, vinblastine, mechloretamine, vincristine, bleomycin, etoposide, and prednisone (Stanford V [StV]) versus the combination of mechlorethamine, vincristine, procarbazine, prednisone (MOPP) with epidoxorubicin, bleomycin, vinblastine (EBV), lomustine
A Paris et al.
Revue des maladies respiratoires, 28(7), 928-932 (2011-09-29)
Pulmonary Langerhans histiocytosis (PLH) is a rare disease due to the accumulation of Langerhans cells at the level of the bronchioles. These dendritic immunocytes form granulomata and destroy the wall of the airway. We report a case of PLH developing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service