Skip to Content
Merck
All Photos(1)

Key Documents

V4877

Sigma-Aldrich

Verrucarin A from Myrothecium sp.

≥85.0% (HPLC)

Synonym(s):

Muconomycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34O9
CAS Number:
Molecular Weight:
502.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥85.0% (HPLC)

storage temp.

−20°C

SMILES string

CC1CCOC(=O)\C=C\C=C\C(=O)OC2CC3OC4C=C(C)CCC4(COC(=O)C1O)C2(C)C35CO5

InChI

1S/C27H34O9/c1-16-8-10-26-14-33-24(31)23(30)17(2)9-11-32-21(28)6-4-5-7-22(29)36-18-13-20(35-19(26)12-16)27(15-34-27)25(18,26)3/h4-7,12,17-20,23,30H,8-11,13-15H2,1-3H3/b6-4+,7-5+

InChI key

NLUGUZJQJYVUHS-YDFGWWAZSA-N

General description

Verrucarin derived from the Myrothecium verrucari, a fungal plant pathogen is a macrocyclic trichothecene compound.

Application

Verrucarin A from Myrothecium sp has been used:
  • as a translation initiation inhibitor in P. falciparum W2 strain
  • as a peptidyl transfer inhibitor to test sensitivity in methyltransferase knock-out S. cerevisiae cells
  • to test its cytotoxicity in renal cell carcinoma cells

Biochem/physiol Actions

Verrucarin A binds to the P and A sites in ribosomes and blocks peptide-bond and translation initiation. It inhibits the clear cell renal cell carcinoma (CCRCC) proliferation. Verrucarin A blocks the peptidyl transferase activity and favors apoptosis induction in cancer cells
Verrucarin A is phytotoxic to plantlet cultures and cytotoxic to cultured mammalian cell lines.

Other Notes

Macrocyclic trichothecene mycotoxin.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antonio L Amelio et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(51), 20314-20319 (2007-12-14)
Signal transduction pathways often use a transcriptional component to mediate adaptive cellular responses. Coactivator proteins function prominently in these pathways as the conduit to the basic transcriptional machinery. Here we present a high-throughput cell-based screening strategy, termed the "coactivator trap,"
Charles S Bond et al.
The Journal of cell biology, 186(5), 637-644 (2009-09-02)
Paraspeckles are ribonucleoprotein bodies found in the interchromatin space of mammalian cell nuclei. These structures play a role in regulating the expression of certain genes in differentiated cells by nuclear retention of RNA. The core paraspeckle proteins (PSF/SFPQ, P54NRB/NONO, and
Chien-Shan Cheng et al.
Cancer cell international, 19, 92-92 (2019-04-24)
In the past decade, increasing research attention investigated the novel therapeutic potential of steroidal cardiac glycosides in cancer treatment. Huachansu and its main active constituent Bufalin have been studied in vitro, in vivo and clinical studies. This review aims to
Girma M Woldemichael et al.
Neoplasia (New York, N.Y.), 14(8), 771-777 (2012-09-07)
Verrucarin A (VA) is a small molecule derived from the fungal plant pathogen Myrothecium verrucaria and was identified as a selective inhibitor of clear cell renal cell carcinoma (CCRCC) cell proliferation in a high-throughput screen of a library of naturally
Dorrah Deeb et al.
International journal of oncology, 49(3), 1139-1147 (2016-08-31)
Pancreatic ductal adenocarcinoma (PDA) remains one of the most difficult to treat of all malignancies. Multimodality regimens provide only short-term symptomatic improvement with minor impact on survival, underscoring the urgent need for novel therapeutics and treatment strategies for PDA. Trichothecenes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service