Skip to Content
Merck
All Photos(1)

Key Documents

V4508

Sigma-Aldrich

Vasoactive Intestinal Peptide Fragment 6-28 human, porcine, rat

≥97% (HPLC)

Synonym(s):

VIP 6-28

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$679.00

$679.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$679.00

About This Item

Empirical Formula (Hill Notation):
C126H207N37O34S
CAS Number:
Molecular Weight:
2816.28
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

$679.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CO)NC(=O)C(CC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CCCCN)NC(=O)C(CCCCN)NC(=O)C(NC(=O)C(C)NC(=O)C(CCSC)NC(=O)C(CCC(N)=O)NC(=O)C(CCCCN)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(CCCNC(N)=N)NC(=O)C(NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(NC(=O)C(N)Cc3ccccc3)C(C)O)C(C)O)C(C)C)C(=O)NC(CC(C)C)C(=O)NC(CC(N)=O)C(N)=O

InChI

1S/C126H207N37O34S/c1-15-66(10)99(122(195)157-86(53-64(6)7)113(186)150-83(102(135)175)57-94(132)170)161-120(193)92(61-164)159-117(190)90(59-96(134)172)155-114(187)85(52-63(4)5)152-115(188)87(55-71-34-38-73(167)39-35-71)153-109(182)77(30-20-23-46-128)144-107(180)78(31-21-24-47-129)148-121(194)98(65(8)9)160-103(176)67(11)142-105(178)82(44-50-198-14)147-111(184)81(42-43-93(131)169)146-106(179)76(29-19-22-45-127)143-108(181)79(32-25-48-140-125(136)137)145-112(185)84(51-62(2)3)151-110(183)80(33-26-49-141-126(138)139)149-123(196)101(69(13)166)163-119(192)88(56-72-36-40-74(168)41-37-72)154-116(189)89(58-95(133)171)156-118(191)91(60-97(173)174)158-124(197)100(68(12)165)162-104(177)75(130)54-70-27-17-16-18-28-70/h16-18,27-28,34-41,62-69,75-92,98-101,164-168H,15,19-26,29-33,42-61,127-130H2,1-14H3,(H2,131,169)(H2,132,170)(H2,133,171)(H2,134,172)(H2,135,175)(H,142,178)(H,143,181)(H,144,180)(H,145,185)(H,146,179)(H,147,184)(H,148,194)(H,149,196)(H,150,186)(H,151,183)(H,152,188)(H,153,182)(H,154,189)(H,155,187)(H,156,191)(H,157,195)(H,158,197)(H,159,190)(H,160,176)(H,161,193)(H,162,177)(H,163,192)(H,173,174)(H4,136,137,140)(H4,138,139,141)

InChI key

BVEZAVADHLXCKB-UHFFFAOYSA-N

Gene Information

human ... VIP(7432)
rat ... Vip(117064)

Amino Acid Sequence

Phe-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2

Biochem/physiol Actions

Potent VIP receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhifang Xu et al.
Scientific reports, 6, 22373-22373 (2016-03-02)
Pituitary adenylate cyclase-activating polypeptide (PACAP, encoded by adcyap1) plays an important role in ectodermal development. However, the involvement of PACAP in the development of other germ layers is still unclear. This study assessed the expression of a PACAP-specific receptor (PAC1)
Fishbein, V.A., et al.
Peptide, 15, 95-95 (1994)
K Shoge et al.
Brain research, 809(1), 127-136 (1998-10-31)
The effects of vasoactive intestinal peptide (VIP) on glutamate-induced delayed death were examined using the primary cultures of rat retinal neurons. Effects of VIP on glutamate-induced neurotoxicity were evaluated by double staining with fluorescein diacetate and propidium iodide. Glutamate (1
Chiara Traini et al.
Neuropeptides, 81, 102031-102031 (2020-03-08)
Glucagon-like peptide-2 (GLP-2) has been reported to indirectly relax gastric smooth muscle. In the present study we investigated, through a combined mechanical and immunohistochemical approach, whether GLP-2 interferes with the electrical field stimulation (EFS)-induced vipergic relaxant responses and the mechanism
R P Mohney et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(14), 5285-5293 (1998-07-03)
Neurons in the adult rat superior cervical sympathetic ganglion (SCG) dramatically increase their content of vasoactive intestinal peptide (VIP) and its mRNA after axotomy in vivo and after explantation. Because the VIP gene contains a functional cAMP response element, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service