V1133
[Arg8, Gly10]-Vasotocin
≥97% (HPLC)
Synonym(s):
Hydrin 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C45H69N15O14S2
CAS Number:
Molecular Weight:
1108.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CCC(C)C1NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(N)CSSCC(NC(=O)C(CC(N)=O)NC(=O)C(CCC(N)=O)NC1=O)C(=O)N3CCCC3C(=O)NC(CCCNC(N)=N)C(=O)NCC(=O)NCC(O)=O
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Chauvet et al.
International journal of peptide and protein research, 45(5), 482-487 (1995-05-01)
From the pituitary neurointermediate lobe of the African toad Bufo regularis, vasotocin, hydrin 2 (vasotocinyl-Gly) and a mesotocin-like peptide have been isolated by HPLC and characterized by mass spectrometry, amino acid sequence and chromatographic coelution with synthetic peptides. The mesotocin-like
J Chauvet et al.
Comparative biochemistry and physiology. Comparative physiology, 104(3), 497-502 (1993-03-01)
1. The neurohypophysial osmoregulatory hormones of the African toad Bufo regularis, a species adapted to estivate under dry and hot conditions, have been investigated. Vasotocin and hydrin 2 (vasotocinyl-Gly) have been identified by their retention times in high-pressure reverse-phase liquid
G Michel et al.
Neuropeptides, 25(2), 139-143 (1993-08-01)
Hydrin 2 (vasotocinyl-Gly) is an intermediate in pro-vasotocin processing found, along with vasotocin, only in the neurohypophysis of anuran amphibians. It increases cutaneous water permeability in the frog and is likely involved in neuroendocrine control of osmoregulation. The relative amounts
J Chauvet et al.
Neuroendocrinology, 51(2), 233-236 (1990-02-01)
Neurohypophyseal secretory granules have been purified from the frog (Rana esculenta) neurointermediate pituitary gland by sucrose gradient centrifugation, and their polypeptide content has been analyzed by reverse-phase high-pressure liquid chromatography. Aside from vasotocin, mesotocin, and their associated neurophysins, hydrin 2
G Michel et al.
FEBS letters, 264(1), 135-137 (1990-05-07)
Hydrin 2 (vasotocinyl-Gly), a hydroosmotic peptide resulting from differential processing of provasotocin and recently identified in frog neurohypophysis, has been looked for in the pituitary gland of an exotic toad (Bufo marinus) and of a reptile (Vipera aspis). Hydrin 2
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service