Skip to Content
Merck
All Photos(1)

Key Documents

U9382

Sigma-Aldrich

Ubiquitin-Carrier Protein H10 human

≥95% (SDS-PAGE), recombinant, expressed in E. coli (histidine-tagged), buffered aqueous glycerol solution

Synonym(s):

UbcH10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$862.00

$862.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$862.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$862.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli (histidine-tagged)

Assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~20 kDa

technique(s)

ligand binding assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBE2C(11065)

General description

Manufactured for Sigma by Boston Biochem., Inc.

Application

Ubiquitin-Carrier Protein H10 (UbcH10) human may be used to study the role of ubiquitin-proteasome system in pathogenesis of human diseases.

Biochem/physiol Actions

In conjunction with anaphase-promoting complex, catalyzes both cyclin A and cyclin B destruction.
In conjunction with the anaphase-promoting complex, the protein catalyzes the destruction of both cyclin A and cyclin B.
Ubiquitin-Carrier Protein H10 human also referred to as UbcH10 is a recombinant protein that catalyzes the destruction of cyclin A as well as cyclin B along with the anaphase promoting complex and hence plays a pivotal role in controlling the cell exit from mitosis.[1]

Physical form

Solution in 50 mM HEPES, pH 8.0, 125 mM NaCl, and 1 mM DTT in 10% glycerol.

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F M Townsley et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(6), 2362-2367 (1997-03-18)
Destruction of mitotic cyclins by ubiquitin-dependent proteolysis is required for cells to complete mitosis and enter interphase of the next cell cycle. In clam eggs, this process is catalyzed by a cyclin-selective ubiquitin carrier protein, E2-C, and the cyclosome/anaphase promoting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service