U9252
Uridine 5′-diphosphate, periodate oxidized sodium salt
80-90%
Synonym(s):
Uridine 5′-diphosphate-2′,3′-dialdehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12N2O12P2
CAS Number:
Molecular Weight:
402.15
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Assay
80-90%
storage temp.
−20°C
SMILES string
[Na].OP(O)(=O)OP(O)(=O)OCC(OC(C=O)N1C=CC(=O)NC1=O)C=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service