Skip to Content
Merck
All Photos(1)

Documents

U2383

Sigma-Aldrich

Ubiquitin Carrier Protein H3/CDC34 human

≥95% (SDS-PAGE), recombinant, expressed in E. coli overproducing strain

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

human

Quality Level

recombinant

expressed in E. coli overproducing strain

Assay

≥95% (SDS-PAGE)

form

solution

mol wt

~32 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

Cdc34 is an essential gene for viability in yeast. Cdc34 mediates the transition from G1 to S-phase of the cell cycle by degrading the S-phase cyclin/CDK inhibitor, SIC1 in yeast and p27(KIP1) in human. Cdc34 is also known to interact with the Skp1/Cdc53/F-box (SCF) ubiquitin ligase subunits Cul1 to degrade the NF-κB inhibitor IκBα in a phosphorylation-dependent manner.
Infected cell protein 0 of herpes simplex virus 1 was shown to bind UbcH3 and induce its degradation by promoting its polyubiquination. This resulted in increased expression of an UbcH3-target gene cyclin D1 and aided in further viral infection.

Physical form

Solution in 50 mM HEPES, 200 mM NaCl, 1 mM TCEP (pH 7.5), and 10% glycerol.

Other Notes

Manufactured for Sigma by Boston Biochem. Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan Hagglund et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(2), 631-636 (2002-01-24)
Infected cell protein 0 (ICP0) of herpes simplex virus 1, a multifunctional ring finger protein, enhances the expression of genes introduced into cells by infection or transfection, interacts with numerous cellular and viral proteins, and is associated with the degradation
Evangelia Koutelou et al.
The Journal of biological chemistry, 283(7), 3846-3853 (2007-12-14)
Notch signaling constitutes an evolutionarily conserved mechanism that mediates cell-cell interactions in various developmental processes. Numerous regulatory proteins interact with the Notch receptor and its ligands and control signaling at multiple levels. Ubiquitination and endocytosis followed by endosomal sorting of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service