Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | $352.00 |
About This Item
NACRES:
NA.71
UNSPSC Code:
12352207
Storage temp.:
−70°C
Shipped in:
dry ice
shipped in
dry ice
storage temp.
−70°C
General description
The gel is formed by stable thioether bonds between thiol groups on CD cell-degradable crosslinker and thio-reactive groups on SLO-PVA. The polymer, SLO-PVA is functionalized with thiol-reactive groups which react with crosslinker at slower rate than maleimides in FAST-PVA. SLO-PVA does not have sites for cell attachment, but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step.
The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala), which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1, MMP3, MMP7 and MMP9).
The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala), which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1, MMP3, MMP7 and MMP9).
Application
TRUE8-1KT is used to generate hydrogels of different stiffness (0.15 to 5 KPa shear modulus) to mimic in-vivo conditions and to investigate the effect of matrix stiffness on cell physiology and behavior. These hydrogels are also employed to study cell migration of neutrophils, dendritic cells and lymphocytes. The SLO-PVA based hydrogels are preferred over dextran-based gels for long-term culture of cells producing carbohydrate degrading enzymes.
Features and Benefits
- Slow gelling (takes 1.5 minutes to 4 minutes, depends on gel stiffness)
- Flexibility in modulating gel stiffness
- Contains MMP-cleavable crosslinker
- Does not allow cell recovery and post-culture analyses
- Cell inert hydrogel (if not customized with RGD peptide)
Other Notes
TRUE8-1KT contains SLO-PVA solution in phosphate buffer and lyophilized CD cell-degradable crosslinker with reconstitution buffer and water.
Disclaimer
Avoid prolonged exposure of CD cell-degradable crosslinker to air and store SLO-PVA in small portions to avoid frequent freeze-thaw cycles.
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. −70°C | storage temp. −70°C | storage temp. −70°C | storage temp. −70°C |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Still not finding the right product?
Explore all of our products under TrueGel3D Hydrogel Kits
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
wgk
WGK 3
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

