Key Documents
TMO342002-9050
Nalgene® sterile diagnostic bottle
high-density polyethylene, bottle capacity 15 mL, natural, case of 896 ea
Synonym(s):
small volume plastic bottle, test kit bottle
Select a Size
896 EACH
$995.00
$995.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
896 EACH
$995.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$995.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
high-density polyethylene
natural
sterility
sterile
packaging
case of 896 ea
manufacturer/tradename
Nalgene 342002-9050
bottle capacity
15 mL
General description
Perform many small volume packing applications with Thermo Scientific Nalgene HDPE Diagnostic Bottles with Closure. Bottles can be used in test kits and are packed in shrink-wrapped SBS trays for easy handling. Trays can be used as an aid for machine fill-line loading. They come assembled with closures without a liner that can wrinkle.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)