Key Documents
TMO3405-42
Nalgene® HDPE Biotainer® Bottles
clear polycarbonate, 5000 mL capacity, case of 6 ea bulk pack
Synonym(s):
PETG bottle, square bottle, sterile bottle
Select a Size
6 EACH
$465.00
$465.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
6 EACH
$465.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$465.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
clear polycarbonate
sterility
sterile
packaging
case of 6 ea bulk pack
manufacturer/tradename
Nalgene 3405-42
capacity
5000 mL
5000 mL capacity
General description
Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific Nalgene PETG Biotainer Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.
Legal Information
Biotainer is a registered trademark of In Vitro Scientific Products
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)