Key Documents
TMO332189-0016
Nalgene® economy wide-mouth bottle
natural high-density polyethylene, volume 500 mL, case of 125 ea
Synonym(s):
HDPE bottle, nalgene HDPE bottle
Select a Size
125 EACH
$506.00
$506.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
125 EACH
$506.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$506.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
natural high-density polyethylene
packaging
case of 125 ea
manufacturer/tradename
Nalgene 332189-0016
capacity
500 mL
volume
500 mL
General description
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)