Skip to Content
Merck
All Photos(1)

Key Documents

TMO331825-BR

Nunc® barcoded latch rack

for 2.0 mL cryogenic tubes, plastic, 42 positions, case of 10 ea

Synonym(s):

tube rack, tube storage

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
$251.00

$251.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025



Select a Size

Change View
10 EACH
$251.00

About This Item

UNSPSC Code:
41122804
NACRES:
NB.12

$251.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


material

plastic

packaging

case of 10 ea

manufacturer/tradename

Nunc 331825-BR

Looking for similar products? Visit Product Comparison Guide

General description

Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific Nunc Universal Latch Rack for Cryogenic Tubes.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service