Key Documents
TMO2403-0250
Nalgene® phthalate-free wash bottle
FEP, bottle volume 250 mL, unlabelled, case of 4 ea
Synonym(s):
fluid diepenser, plastic wash bottle
Select a Size
4 EACH
$1,280.00
$1,280.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EACH
$1,280.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$1,280.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
FEP
packaging
case of 4 ea
manufacturer/tradename
Nalgene 2403-0250
bottle volume
250 mL
General description
Ensure contamination-free dispensing with Thermo Scientific Nalgene Wash Bottles made with PTFE FEP, useful for trace element work. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymers. Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)