Key Documents
TMO2401-0500
Nalgene® economy wash bottle
low-density polyethylene, volume 500 mL, unlabelled, case of 24 ea
Synonym(s):
LDPE flush bottle, LDPE wash bottle
Select a Size
24 EACH
$226.00
$226.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
24 EACH
$226.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$226.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
low-density polyethylene
packaging
case of 24 ea
manufacturer/tradename
Nalgene 2401-0500
capacity
500 mL
volume
500 mL
General description
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)