Key Documents
TMO2401-0250
Nalgene® economy wash bottle
low-density polyethylene, volume 250 mL, unlabelled, case of 36 ea
Synonym(s):
LDPE flush bottle, LDPE wash bottle
Select a Size
36 EACH
$281.00
$281.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
36 EACH
$281.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$281.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
low-density polyethylene
packaging
case of 36 ea
manufacturer/tradename
Nalgene 2401-0250
capacity
250 mL
volume
250 mL
General description
Prevent separation when squeezing Thermo Scientific Nalgene LDPE Economy Wash Bottles, featuring a tight-fitting draw tube and closure. These low density polyethylene bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. Cut back tip to increase flow.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)