Key Documents
TMO2123-0010
Nalgene® bottles, style 2123
natural high-density polyethylene, volume 4000 mL, case of 6 ea
Synonym(s):
nalgene bottle, nalgene bottles
Select a Size
6 EACH
$322.00
$322.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
6 EACH
$322.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$322.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
natural high-density polyethylene
polypropylene copolymer bottle (PPCO)
description
Autoclavable
packaging
case of 6 ea
manufacturer/tradename
Nalgene 2123-0010
capacity
4000 mL
volume
4000 mL
General description
This wide-mouth polypropylene copolymer bottle with closure is excellent for collecting, storing and transporting dry chemicals and environmental samples. Square, space saving design features molded-in handgrips for greater convenience and safety when pouring, autoclavable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)