Key Documents
TMO161093
Nunc® MicroWell 96-well Plate with high flange skirt
Nunclon® Delta surface, polystyrene, sterile, with lid, clear, case of 160 ea
Synonym(s):
multiwell plate, treated multiwell plate, treated plate
Select a Size
160 EACH
$1,230.00
$1,230.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
160 EACH
$1,230.00
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
$1,230.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
clear
polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 161093
General description
Obtain consistent and reliable results with Thermo Scientific Nunc MicroWell 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)