Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 160 ea | Check Cart for Availability | $1,530.00 |
About This Item
$1,530.00
material
polystyrene , white
sterility
sterile
feature
lid, pinchbar design
packaging
case of 160 ea
manufacturer/tradename
Nunc 136102
plate L × W
128 mm × 86 mm
size
96 wells
well volume
(400 μL)
working volume
250 μL
suitability
suitable for (fluorescence-based cell assays)
General description
Features and Benefits
- White plates provide maximum reflection, minimum auto-fluorescence and autoluminescence.
- Black plates provide minimal background and light scatter in fluorescence assays.
- Non-toxic white or black plate is provided with clear lid.
- Raised well rims of flat bottom wells reduce risk of cross contamination.
- Low crosstalk.
- Nunclon™ Δ for cell based assays.
- Working volume range: 50-250 μl/well.
- Suitable for fluorescence-based cell assays.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| feature lid, pinchbar design | feature lid, pinchbar design | feature lid, pinchbar design | feature lid, pinchbar design |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Nunc 136102 | manufacturer/tradename Nunc 137103 | manufacturer/tradename Nunc 136101 | manufacturer/tradename Nunc 137101 |
| material polystyrene , white | material black , polystyrene | material flat bottom white polystyrene wells | material black , flat bottom black polystyrene wells |
| suitability suitable for (fluorescence-based cell assays) | suitability suitable for (fluorescence-based cell assays) | suitability suitable for (fluorescence-based cell assays) | suitability suitable for (fluorescence-based cell assays) |
| working volume 250 μL | working volume 250 μL | working volume 250 μL | working volume 250 μL |
Still not finding the right product?
Explore all of our products under Nunc® MicroWell™ 96 well polystyrene plates
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



