Key Documents
TMO132920
Nunc® TripleFlasks, Nunclon® Δ Surface
Nunclon® Delta, surface area (500 cm2), filter cap, straight neck, sterile, case of 32 ea
Synonym(s):
cell culture flask, tissue culture flask
Select a Size
32 EACH
$1,020.00
$1,020.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
32 EACH
$1,020.00
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
$1,020.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
screw top blue high-density polyethylene cap
straight neck
straight neck transparent polystyrene flask
sterility
sterile
sterile
packaging
case of 32 ea
manufacturer/tradename
Nunc 132920
flask working volume
200 mL
surface area
(500 cm2)
500 cm2 , culture area (cm2) per flask
working volume
200 mL
binding type
Nunclon Delta surface
General description
Perform cell culture expansion and experience 500cm2 total growth area within the footprint of a standard flask using the Nunc® TripleFlask™ Treated Cell Culture Flasks. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. Three layers of growth surfaces provide a total culture area of 500 cm2 in the same footprint as a standard Nunc 175cm2 flask.
Features and Benefits
  • Three parallel growth surfaces provide a total culture area of 500 cm2
  • External dimensions of a 175 cm2 standard flask
  • Ideal for scale-up cell culture applications
  • Extra caps in every carton
  • Nunclon™ Δ surface ensures consistent optimal cell attachment and growth
  • Barcoded with large Code 128
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)