Skip to Content
Merck
All Photos(1)

Key Documents

T9265

Sigma-Aldrich

Thyrotropic hormone from human pituitary

lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)

Synonym(s):

hTSH, TSH, Thyroid stimulating hormone

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$244.00
10 μG
$428.00
25 μG
$856.00
0.1 MG
$2,830.00

$244.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$244.00
10 μG
$428.00
25 μG
$856.00
0.1 MG
$2,830.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$244.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

biological source

human pituitary glands

Quality Level

sterility

non-sterile

form

lyophilized powder

specific activity

≥6.2 IU/mg (immunoassay, non-sterile)

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

technique(s)

cell culture | mammalian: suitable

impurities

HIV Antibodies
Antibody to HCV
Antibody to HBV or Presence of HBSAG, tested negative

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Thyrotropic hormone or thyroid stimulating hormone (TSH) belongs to the glycoprotein hormone family.[1] Thyrotroph cells of the anterior pituitary gland produces TSH and is stored in the secretory granules. It comprises a heterodimer of two noncovalently linked subunits, α and β.[2]

Application

Thyrotropic hormone from human pituitary has been used as a supplement in the fetal thyroid organoid culture medium for stimulating maturation.[3] It has also been used as a medium supplement for primary cell culture, and is used further to study the molecular mechanisms of estrogen-mediated growth stimulation of human thyroid tumor cells.[4]

Biochem/physiol Actions

In children, thyroid hormones are very much essential for the normal cognition and neurodevelopment.[5] Thyroid-stimulating hormone (TSH) test is mainly used to diagnose thyroid function.[6]
The product can enhance vascular endothelial growth factor (VEGF) secretion and angiogenesis in thyroid cancer celllines.[7]TSHR activated by TSH activates both the adenosine 3′,5′-cyclic monophosphate (cAMP) pathway (via Gsa) and the phosphoinositol/calcium (IP/Ca2+; via Gq) second messenger signaling pathway by interacting with Gs and Gq G-proteins.[8] In mature adipocytes, TSH aids in lipid metabolism by inhibiting fatty acid synthase (FASN) expression via, activating protein kinase A (PKA)-dependent phosphorylation of extracellular signal-regulated kinases/p42/p44 (ERK1/2), and c-Jun N-terminal kinase (JNK).[9]
Two-chain glycoprotein hormone. The α-chain is not active, biological specificity attributed rather to the β-chain. Activates adenylate cyclase in the thyroid gland, stimulating iodine uptake, thyroxine synthesis and release. Goitrogenic.

Packaging

Package size based on hormone

Physical form

Contains hormone and salts from approx. 0.1 mL of 0.05 M phosphate buffer, pH 7.4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thyroid-stimulating hormone
The pituitary, 163-201 (2017)
D Manole et al.
The Journal of clinical endocrinology and metabolism, 86(3), 1072-1077 (2001-03-10)
Thyroid tumors are about 3 times more frequent in females than in males. Epidemiological studies suggest that the use of estrogens may contribute to the pathogenesis of thyroid tumors. In a very recent study a direct growth stimulatory effect of
Impact of light exposure on thyroid-stimulating hormone results using the Siemens Advia Centaur TSH-3Ultra assay
Armer J, et al.
Annals of Clinical Biochemistry, 54(5), 612-615 (2017)
Thyroid-stimulating hormone and thyroid-stimulating hormone receptor structure-function relationships
Szkudlinski MW, et al.
Physiological Reviews, 82(2), 473-502 (2002)
Zhenghao Wu et al.
Journal for immunotherapy of cancer, 10(1) (2022-02-02)
Hormones are identified as key biological variables in tumor immunity. However, previous researches mainly focused on the immune effect of steroid hormones, while the roles that thyroid-stimulating hormone (TSH) played in the antitumor response were far from clear. The source

Questions

  1. How to reconstitute this product.

    1 answer
    1. This product may be reconstituted in 0.01M phosphate buffer at pH 7.5. Stock solutions, prepared at 10 micro IU per milliliter, may be stored frozen in aliquots. Avoid repeated freeze/thaw cycles. Working solutions should be prepared fresh and not stored.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service