Skip to Content
Merck
All Photos(2)

Key Documents

T8902

Sigma-Aldrich

Thiostrepton from Streptomyces azureus

from Streptomyces azureus, ≥90% (HPLC), powder, FoxM1 inhibitor

Synonym(s):

Bryamycin, NSC 170365, NSC 81722, Thiactin, Thiostreptin A

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$208.00

$208.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$208.00

About This Item

Empirical Formula (Hill Notation):
C72H85N19O18S5
CAS Number:
Molecular Weight:
1664.89
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$208.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Thiostrepton from Streptomyces azureus, ≥90% (HPLC)

Assay

≥90% (HPLC)

solubility

acetic acid: soluble 25 mg/mL, clear to hazy, yellow to brownish-yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
parasites

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CCC(C)C1NC2C=Cc3c(cc(nc3C2O)C(=O)OC(C)C4NC(=O)c5csc(n5)C(NC(=O)C6CSC(=N6)C(\NC(=O)C(NC(=O)c7csc(n7)C8(CCC(=NC8c9csc4n9)c%10nc(cs%10)C(=O)NC(=C)C(=O)NC(=C)C(N)=O)NC(=O)C(C)NC(=O)C(=C)NC(=O)C(C)NC1=O)C(C)O)=C\C)C(C)(O)C(C)O)C(C)O

InChI

1S/C72H85N19O18S5/c1-14-26(3)47-63(105)78-30(7)57(99)75-28(5)56(98)76-31(8)58(100)91-72-19-18-40(66-85-43(22-111-66)59(101)77-29(6)55(97)74-27(4)54(73)96)81-52(72)42-21-112-67(83-42)49(34(11)109-69(107)41-20-37(32(9)92)36-16-17-39(79-47)51(95)50(36)80-41)89-60(102)44-24-113-68(86-44)53(71(13,108)35(12)94)90-62(104)45-23-110-65(84-45)38(15-2)82-64(106)48(33(10)93)88-61(103)46-25-114-70(72)87-46/h15-17,20-22,24-26,30-35,39,45,47-49,51-53,79,92-95,108H,4-6,14,18-19,23H2,1-3,7-13H3,(H2,73,96)(H,74,97)(H,75,99)(H,76,98)(H,77,101)(H,78,105)(H,82,106)(H,88,103)(H,89,102)(H,90,104)(H,91,100)/b38-15-

InChI key

NSFFHOGKXHRQEW-BXVAPQLOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Thiostrepton has been used as a selectable marker for recombinant Streptomyces cultures[1],[2],[3].

Biochem/physiol Actions

Peptide antibiotic that prevents the binding of elongation factor G (EF-G) and GTP to the 50S ribosomal subunit.

Preparation Note

Thiostrepton dissolves in acetic acid at 25 mg/ml to yield a clear to hazy, yellow-yellow brown solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M V Rodnina et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(17), 9586-9590 (1999-08-18)
The region around position 1067 in domain II of 23S rRNA frequently is referred to as the GTPase center of the ribosome. The notion is based on the observation that the binding of the antibiotic thiostrepton to this region inhibited
Feifei Zhang et al.
Methods in enzymology, 516, 3-24 (2012-10-05)
Thiopeptides are a family of highly modified peptide metabolites, characterized by a macrocycle bearing a central piperidine/dehydropiperidine/pyridine ring, multiple thiazole rings, and several dehydrated amino acid residues. Thiopeptides have useful antibacterial, antimalarial, and anticancer properties but have not been adapted
Antje Wolf et al.
Journal of molecular modeling, 19(2), 539-549 (2012-09-11)
With improvements in computer speed and algorithm efficiency, MD simulations are sampling larger amounts of molecular and biomolecular conformations. Being able to qualitatively and quantitatively sift these conformations into meaningful groups is a difficult and important task, especially when considering
M L Chiu et al.
The Journal of biological chemistry, 274(29), 20578-20586 (1999-07-10)
Microbial metabolites isolated in screening programs for their ability to activate transcription of the tipA promoter (ptipA) in Streptomyces lividans define a class of cyclic thiopeptide antibiotics having dehydroalanine side chains ("tails"). Here we show that such compounds of heterogeneous
Takamichi Ito et al.
Scientific reports, 14(1), 4048-4048 (2024-02-20)
Extramammary Paget disease (EMPD) is a rare skin cancer that primarily affects older individuals predominantly in areas with apocrine sweat glands. Although most early EMPD lesions are indolent, patients with metastatic EMPD have a poor prognosis due to the lack

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service