Skip to Content
Merck
All Photos(2)

Key Documents

T8375

Sigma-Aldrich

DL-Threonine

≥95% (titration)

Synonym(s):

(±)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$453.00

$453.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$453.00

About This Item

Linear Formula:
CH3CH(OH)CH(NH2)COOH
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721647
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$453.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Product Name

DL-Threonine, ≥95% (titration)

Assay

≥95% (titration)

form

powder

impurities

≤30% allo-threonine

color

white to off-white

mp

244 °C (dec.) (lit.)

SMILES string

C[C@H](O)[C@@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m0/s1

InChI key

AYFVYJQAPQTCCC-STHAYSLISA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Gimesia chilikensis sp. nov., a haloalkali-tolerant planctomycete isolated from Chilika lagoon and emended description of the genus Gimesia.: The study describes the isolation and characterization of a new bacterial species using ᴅʟ-Threonine as part of the nutrient medium, demonstrating its role in microbial ecology research (Kumar et al., 2020).


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pradeep K Das Mohapatra et al.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi, 42(2), 172-175 (2009-07-15)
Microbial tannase (tannin acyl hydrolase, EC 3.1.1.20), a hydrolysable tannin-degrading enzyme, has gained importance in various industrial processes, and is used extensively in the manufacture of instant tea, beer, wine, and gallic acid. Tannase is an inducible enzyme, and hydrolysable
Guo-qiang Zhang et al.
Analytical biochemistry, 409(1), 130-137 (2010-10-19)
Bacillus amyloliquefaciens has been a major workhorse for the production of a variety of commercially important enzymes and metabolites for the past decades. Some subspecies of this bacterium are recalcitrant to exogenous DNA, and transformation with plasmid DNA is usually
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service