Skip to Content
Merck
All Photos(1)

Key Documents

T8325

Sigma-Aldrich

TDZD-8

≥98% (HPLC), needles

Synonym(s):

4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$225.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2O2S
CAS Number:
Molecular Weight:
222.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

needles

storage condition

protect from light

color

white

mp

63-64.4 °C

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

CN1SC(=O)N(Cc2ccccc2)C1=O

InChI

1S/C10H10N2O2S/c1-11-9(13)12(10(14)15-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

JDSJDASOXWCHPN-UHFFFAOYSA-N

Application

TDZD-8 has been used to study the role of GSK-3 in maintaining MLL leukemia stem cell transcriptional program[1]. TDZD-8 has also been used to analyze its effect on neural functions of the mouse olfactory bulb[2].

Biochem/physiol Actions

TDZD-8 is a selective inhibitor of GSK-3, a thiadiazolidinone derivative, non-ATP competitive inhibitor of GSK-3β (IC50 = 2 μM). It does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. TDZD-8 has been proposed to bind to the kinase site of GSK-3β.
TDZD-8 is a selective inhibitor of GSK-3.

Features and Benefits

This compound is featured on the GSK-3 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

TDZD-8 is soluble in DMSO at a concentration that is greater than 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qing Yu et al.
Cell death & disease, 15(1), 73-73 (2024-01-19)
Chronic stress and elevated levels of glucocorticoids (GCs), the main stress hormones, accelerate Alzheimer's disease (AD) onset and progression. A major driver of AD progression is the spreading of pathogenic Tau protein between brain regions, precipitated by neuronal Tau secretion.
Teng Jiang et al.
Inflammation, 41(3), 811-823 (2018-01-25)
As a recently identified susceptibility gene for Alzheimer's disease (AD), triggering receptor expressed on myeloid cells 2 (TREM2) encodes an immune receptor that is uniquely expressed on microglia, functioning as a modulator of microglial functions including phagocytosis and inflammatory response.
Jun Yoshida et al.
Lipids, 52(3), 295-301 (2017-02-15)
Many uncommon non-methylene-interrupted fatty acids (NMI FA) are present in limpet gonads, but their biological properties remain unknown. To investigate new biological effects of naturally occurring NMI FA in eukaryotic cells, the biological activities of structurally analogous (4Z,15Z)-octadecadienoic acid (1)
Bo Xing et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 41(13), 3003-3015 (2016-06-30)
Schizophrenia (SZ) is a neurodevelopmental disorder in which the emergence of cognitive symptoms occurs during early adolescence. Glycogen synthase kinase-3β (GSK3β) plays a critical role in synaptic plasticity during development and is highly implicated in the etiology of SZ. However
Lydia Hannou et al.
Journal of biological rhythms, 33(2), 166-178 (2018-04-20)
NEUROLIGIN-1 (NLGN1) is a postsynaptic adhesion molecule involved in the regulation of glutamatergic transmission. It has been associated with several features of sleep and psychiatric disorders. Our previous work suggested that transcription of the Nlgn1 gene could be regulated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service