Skip to Content
Merck
All Photos(3)

Key Documents

T8203

Sigma-Aldrich

Anti-TUBA1A (TUBA3) Antibody

enhanced validation
Anti-α-Tubulin antibody, Mouse monoclonal

mouse monoclonal, AA13

Synonym(s):

Anti-Tuba1a, Anti-Tubulin, α1A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AA13, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~2.0 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using Neuro-2a cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)
mouse ... Tuba1a(22142)
rat ... Tuba1a(64158)

General description

Anti-α-Tubulin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma AA13 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with rat brain tubulin.
Tubulin α 1a (TUBA1A) is a member of α-tubulin gene family. It is encoded by the gene mapped to human chromosome 12q13.12. Tubulin α 1a is a vital structural subunit of microtubules that are temporarily expressed in almost all post-mitotic neurons during neuronal development.
The encoded protein structure contains core of two β-sheets enclosed by α-helices. TUBA1A is characterized with an amino-terminal domain with the guanine nucleotide-binding region, an intermediate domain associated with interprotofilament lateral contacts and the modulation of tubulin curvature, and a carboxy-terminal domain that contains region involved in interacting with microtubule-associated proteins (MAPs) and molecular motors, such as kinesins and dynein.

Immunogen

rat brain tubulin.

Application

Anti-α-Tubulin antibody, Mouse monoclonal has been used in western blotting.
The antibody has been used in various immunochemical techniques including immunoprecipitation, immunocytochemistry, and immunohistochemistry.

Biochem/physiol Actions

Gene mutation of tubulin α-1A (TUBA1A) is observed in perisylvian asymmetrical polymicrogyria, polymicrogyria-like cortical dysplasia, and microlissencephaly in foetal cases. Alteration in the expression of the gene impairs neural migration and causes lissencephaly (LIS).
Tubulin is the major building block of microtubules, which is a structural and mobile element in mitosis, intracellular transport, flagellar movement and the cytoskeleton. Monoclonal antibodies recognizing α-tubulin, together with monoclonal antibodies to other tubulin types (α-tubulin isotypes I, II, and III, tyrosine tubulin and acetylated- α -tubulin) provide a specific and useful tool in studying the intracellular distribution of tubulin, and the static and dynamic aspects of the cytoskeleton. In addition, the various tubulin forms have been implicated in cancerous processes and are thus suggested as potential targets for cancer therapy.

Target description

Tubulin is a cylindrical, filamentous structure is present inalmost all eukaryotic cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New tubulin targeting agents currently in clinical development
Carlson RO
Expert Opinion on Investigational Drugs, 17(5), 707-722 (2008)
Anaphase-promoting complex/cyclosome participates in the acute response to protein-damaging stress.
Ahlskog JK
Molecular and Cellular Biology, 30(24), 5608-5620 (2010)
TUBA1A mutation can cause a hydranencephaly-like severe form of cortical dysgenesis.
Yokoi S
Scientific Reports, 5:15165 (2015)
Expanding the spectrum of TUBA1A-related cortical dysgenesis to Polymicrogyria.
Poirier K
European Journal of Human Genetics, 21(4), 381-385 (2013)
A case of TUBA1A mutation presenting with lissencephaly and Hirschsprung disease.
Brain & Development, 36(2), 159-162 (2014)

Questions

  1. I'm using this antibody and I'd be interested to know what the region is of peptides that react antibody to alpha-tubulin (T8203) and whether the C-terminal acid domain is involved.

    1 answer
    1. The immunogen used to make T8203 was purified tubulin from rat brain. The epitope to which clone AA13 binds has not been mapped.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service